Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G3 Dialysis Cassettes, 3.5K MWCO
GREENER_CHOICE

Catalog No. PIA52967
Change view
Click to view available options
Capacity (Metric):
3 mL
15 mL
30 mL
70 mL
125 mL
Quantity:
10 cassettes
6 cassettes
8 cassettes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
PIA52967 15 mL 8 cassettes
PIA52966 3 mL 10 cassettes
PIA52968 30 mL 6 cassettes
PIA52969 70 mL 6 cassettes
PIA52970 125 mL 6 cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. PIA52967 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' A52967
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Thermo Scientific Slide-A-Lyzer™ G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 0.5 to 125 mL. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.

The Slide-A-Lyzer G3 Cassettes are single-use dialysis devices designed to maximize performance and practicality. They provide an easy-to-use and secure alternative to traditional dialysis tubing and are used in a wide range of applications including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.

The Slide-A-Lyzer G3 Cassettes are single-use dialysis devices designed to maximize performance and practicality. They provide an easy-to-use and secure alternative to traditional dialysis tubing and are used in a wide range of applications including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.

The new G3 cassettes are ergonomically designed to be significantly lighter and more compact than the previous generations, making them easier to handle and more practical to use. This new design offers a new cap allowing for effortless sample loading and retrieval providing visual and haptic feedback upon opening and closure to assure sample transfer and security. Sample loading and removal is easily accomplished using a serological pipette or hypodermic needle (optional) attached to a syringe. All G3 cassettes offer a built-in air chamber that provides buoyancy and vertical orientation during the dialysis process.

Slide-A-Lyzer G3 Dialysis Cassettes are available in five different sizes, accommodating sample volumes ranging from 0.5 to 125 mL, and are provided with 2K, 3.5K, 10K, and 20K molecular weight cut offs (MWCOs).

The cassettes’ membranes are composed of low-binding regenerated cellulose that provides maximum sample purity and recovery. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

The Slide-A-Lyzer G3 Cassettes with the 10K MWCO are also offered in the Electron Beam treated format for applications such as culturing cells, purifying viruses, or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. A dosage that has been determined to sterilize units was used based on bioburden testing for identifying anaerobic growth. Sterility testing is not performed on individual lots.

Features of Slide-A-Lyzer G3 Dialysis Cassettes include:
Efficient Dialysis—flat cassette chamber, housing two membranes, provides high surface-area to volume ratio maximizing diffusion rate
Ease-of-use—sample loading and removal with a pipette and/or syringe
High sample recovery—low-binding plastic and membrane help minimize sample loss compared to filtration and resin systems
Maximum sample security—sealed membranes prevent leakage that can occur with dialysis tubing and homemade devices. Caps have visual and haptic feedback assuring secure closure of the sample chamber
Stream-lined design—less plastic, lighter weight devices
Self-floating format—designed to float independently without the use of external buoys
Irradiated option—E-beam irradiated cassettes are available for applications requiring conditions that minimize risk of sample contamination

TRUSTED_SUSTAINABILITY

Specifications

Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
Capacity (Metric) 15 mL
MWCO 3.5 kDa
Product Line Slide-A-Lyzer
Quantity 8 cassettes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.