Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Inhibitory Mold Agar
GSA_VA

Catalog No. R01505
Change view
Click to view available options
Quantity:
10 x 26 mL
10 x 35 mL
100/Pk.
Format:
20mm x 113mm Tube
Monoplate - 26mL
Monoplate - 35mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01505 10 x 26 mL Monoplate - 26mL
R01508 10 x 35 mL Monoplate - 35mL
R08410 100/Pk. 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01505 Supplier Thermo Scientific™ Supplier No. R01505
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Primarily isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi using Inhibitory Mold Agar (w/chloramphenicol).

Thermo Scientific™ Remel™ Inhibitory Mold Agar (w/chloramphenicol) is used to primarily isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi. Ulrich formulated Inhibitory Mold Agar as a general-purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, H. capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3.

Inhibitory Mold Agar primarily supports isolation and cultivation of dermatophytes and other pathogenic fungi. Inhibitory Mold Agar contains casein and meat peptones that supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts are essential electrolytes that help to maintains osmotic equilibrium. Yeast extract is a source of vitamins.

  • Ready to use - Solid media
  • Recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3
  • Easy to read - Examine plates for fungal colonies that exhibit typical color and morphology
  • Selective media - Chloramphenicol inhibits many gram-positive and gram-negative bacteria while gentamicin, ciprofloxacin, and vancomycin inhibit commensal microbial flora

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Inhibitory Mold Agar
Format Monoplate - 26mL
Product Type Agar Plate
Quantity 10 x 26 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.