Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ™ Remel™ Anaerobic Reducible LKV Blood Agar/ Bacteroides Fragilis Isolation Agar

Catalog No. R02006
Change view
Click to view available options
Certifications/Compliance:
USDA, APHIS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
R02006 USDA, APHIS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R02006 Supplier Thermo Scientific™ Supplier No. R02006
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Anaerobic Reducible LKV Blood Agar/ Bacteroides Fragilis Isolation Agar (BBE)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USDA, APHIS
Manufacturing Origin United States
Media Type Anaerobic reducible LKV Blood Agar/Bacteroides Fragilis isolation Agar
Formulation Gentamicin <0.1%, Kanamycin <0.1% and Vancomycin <0.1%
For Use With (Application) Isolation and differentiation of microorganisms from samples or specimens
Packaging Type Biplate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.