Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Gentamicin
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective, ready-to-use medium.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) prepared media plates. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concertation of dextrose from 40 to 20 g/L2. The addition of antibiotics in the medium makes the medium inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi.
- Chloramphenicol inhibits some gram-positive and gram-negative bacteria
- Gentamicin inhibits some gram-negative bacteria
Specifications
Specifications
| Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Gentamicin |
| Format | 20mm x 113mm Tube |
| Product Type | Agar Plate |
| Quantity | 20/Pk. |