Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis

Catalog No. R30166801
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

TRUSTED_SUSTAINABILITY

Specifications

Description Group A
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Organism Group Group A
Target Organism Class Neisseria Meningitidis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.