Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Chemical Equations Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28486 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28486 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including soluble Nickel Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28486 Supplier Innovating Science™ Supplier No. IS2514
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including soluble Nickel Compounds, which are known to the State of California to cause cancer and reproductive harm.

To perform a series of experiments highlighting the differences in two common types of chemical reactions; single replacement and double replacement. Innovating Science Introduction To Chemical Equations Educational Kit, help students to identify chemical reactions and write balanced chemical reaction equations.

  • Contains enough materials for 15 groups
  • Teacher's manual and Student Study Guide copymasters are included
  • Kit Includes:
    • 0.1M Aluminum Nitrate (60 mL)
    • 0.1M Cobalt Nitrate (60 mL)
    • 0.1M Cupric Nitrate (60 mL)
    • 0.1M Ferric Nitrate (60 mL)
    • 0.1M Nickel Nitrate (60 mL)
    • 0.1M Silver Nitrate (150 mL)
    • 0.2M Sodium Hydroxide (300 mL)
    • 2.0M Hydrochloric Acid (150 mL)
    • 6 g Zinc Metal
    • 15 pieces Copper Wire
    • 1 pack Wooden Dowels

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1 Kit
Dimensions (L x W x H) 13 x 10 x 7 in.
Class Size 15 Lab Groups
Product Type Introduction to Chemical Equations Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.