Learn More
Description
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
- Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
- Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
- Sterilization of Aqueous Solutions (150 mL to 1 L)
- Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Specifications
Specifications
| Color | Clear with Blue Adapter |
| Filter Type | Durapore |
| Material | Polystyrene |
| Material (Membrane) | PVDF |
| Pore Size | 0.22 μm |
| Sterility | Sterile |
| No. per Pack | 12 |
| Outlet Fitting | 45 mm |
| Sterilization Method | Radio sterilized |
| Volume (Metric) | 500 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.