Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfur Trioxide Pyridine Complex

Catalog No. S755625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
25 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S755625G 1 g
S7556100G 1 kg
S75561KG 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. S755625G Supplier Sigma Aldrich Supplier No. S755625G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sulfur trioxide-pyridine (SO3-py) complex can be used to convert the allylic or benzylic hydroxyl group of compounds such as geraniol, farnesol, and 1-indanol into sulfate monoester, which is a good leaving group. This method is highly useful in the total synthesis of sesquiterpenes.As a sulfating agent, SO3-py complex can be used for the preparation of sulfated-chitins, sulfated hyaluronan (HyaS) derivatives and sulfated starch.

Specifications

CAS 8414-5-8-9
Molecular Formula C14H14O2Si
Linear Formula C14H14O2Si
Quantity 1 g
Synonym NSC 75831; Pyridine sulfur trioxide complex
Molecular Weight (g/mol) 242.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.