Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Acids, Bases and the pH Scale
GREENER_CHOICESCI_ED

Catalog No. S96406 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S96406 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96406 Supplier Innovating Science™ Supplier No. IS2507
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 25mL each of deionized water, dilute acid (0.1M HCl), dilute base (0.1M NaOH), 0.5% litmus, 0.02% Methyl Red, 0.5% Bromothymol Blue, 1.0% Phenolphthalein, vinegar, household ammonia, soap solution, filtered water, Vitamin C solution, unknown solution #1 (water), unknown solution #2 (dilute base), unknown solution #3 (phenolphthalein) and unknown solution #4 (dilute acid); wide-range pH test strips (pack of 50); reaction trays (15); teacher's guide; and copy-masters

Learn about the properties of acids and bases, the role of hydrogen and hydroxide ions and the nature of the pH scale.

  • Examine the effects of acids and bases on several chemical pH indicators
  • Determine the pH of common household materials
  • Use the knowledge gained to determine the composition of four unknown clear solutions

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To learn about the properties of acids and bases, the role of hydrogen and hydroxide ions and the nature of the ph scale
Includes 25mL Each of deionized water, dilute acid (0.1M HCl), dilute base (0.1M NaOH), 0.5% litmus, 0.02% Methyl Red, 0.5% Bromothymol Blue, 1.0% Phenolphthalein, Vinegar, household Ammonia, soap solution, filtered water, Vitamin C solution, unknown solution #1 (water), unknown solution #2 (dilute base), unknown solution #3 (phenolphthalein) and unknown solution #4 (dilute acid), wide-range pH test strips, 15 reaction trays, teacher's guide, and copy-maste
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Acids, Bases, and the pH Scale Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.