Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7.00 (Certified), Fisher Chemical™

Catalog No. SB10820 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Poly Bottle
PolyPac™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB108-20 20 L PolyPac™
SB108-500 500 mL Poly Bottle
SB108-1 1 L Poly Bottle
SB108-10 10 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SB108-20
Add to Cart

Potassium Phosphate Monobasic/Sodium Hydroxide

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 7.00
Melting Point 0°C
CAS 7778-77-0, 1310-73-2, 7732-18-5
Color Colorless
Density 1.003g/cm³
pH 7.00
Physical Form Liquid
Quantity 20 L
Identification Pass Test
MDL Number MFCD00134130
Packaging PolyPac™
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg at 20°C
Synonym Chlorine Titrimeter Reagent
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.