Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ SterilElite™ Vertical Autoclave

Catalog No. STE50VAR Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
50 L
82 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
STE50VAR 50 L
STE82VMB 82 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STE50VAR
Add to Cart
Add to Cart
Request a Quote

Vertical advanced autoclave includes vacuum and dry function

  • Door Mechanism: Hand wheel type of quick-open door with safety lock system
  • Control System: Micro-computer controlled with LCD display and touch-type keys
  • Independent Temperature and Pressure Monitoring: Ensures precise control during sterilization processes
  • High-Precision PT100 Sensor: Accurately controls liquid temperature
  • Built-in Steam Generator: Provides efficient steam production for sterilization
  • Overheating and Pressure Overload Protection: Enhances safety during operation

Specifications

Autoclavable Autoclavable
Certifications/Compliance Quality Management System
Color Blue
Capacity (English) 13.2 gal.
Capacity (Metric) 50 L
Depth (Metric) Chamber 710 mm
Diameter (English) Chamber 11.8 in.
Depth (English) Chamber 28 in.
Diameter (Metric) Chamber 300 mm
For Use With (Application) Decontamination of Biohazardous Waste
For Use With (Equipment) Sterilization of Culture Media
Length (English) 24 in.
Length (Metric) 600 mm
Product Type Autoclave
Temperature (English) 249°F to 275°F
Temperature (Metric) 121°C to 135°C
Weight (English) 178 lb.
Weight (Metric) 81 kg
Includes 2 Stainless Steel Baskets, Kinclean Chamber and Piping Cleaning Agents
Material Stainless Steel
Latex Free Yes
Voltage 230 V
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.