Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetramethylammonium Tetrafluoroborate

Catalog No. T1955025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1955025G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1955025G Supplier Sigma Aldrich Supplier No. T1955025G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetramethylammonium Tetrafluoroborate

Specifications

CAS 107-20-0
Molecular Formula C80H159NO40
Linear Formula C80H159NO40
Quantity 100 mg
MDL Number MFCD21363214
Synonym t-Boc-N-amido-PEG36-acid; t-Boc-PEG-acid; Boc-NH-PEG36-COOH; Boc-NH-PEG36-acid; BocNH-PEG1500-acid
Molecular Weight (g/mol) 1775.1 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.