Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Terphenyl

Catalog No. T3009100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3009100G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3009100G Supplier Sigma Aldrich Supplier No. T3009100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromopropiophenone (α-bromopropiophenone) is an α-bromoketone
  • It participates in the manufacturing process of 1-acetamido-3-methyl-2-phenylimidazo[1,2-a]pyridinium bromide.2-bromopropiophenone (α-bromopropiophenone) may be used for the synthesis of 2-phenylmorpholinols
  • It may also be used in the synthesis of n-phenacylammonium salts [n-(α-benzoylbenzyl)-, n-(1-benzoylethyl)-, n-phenacyl-, pyrazinium, 3-bromoquinolinium, benzothiazolium, or p-cyanopyridinium hexafluoroantimonates]

Specifications

CAS 92-06-8
Melting Point 84°C to 88°C (lit.)
Boiling Point 379°C (lit.)
Molecular Formula C18H14
Linear Formula C6H5C6H4C6H5
MDL Number MFCD00003059
Quantity 25 mL
Synonym 1,3-Diphenylbenzene
Molecular Weight (g/mol) 230.3
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.