Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl Phosphite

Catalog No. T61204100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T61204100ML 100 mL
T612045ML 5 mL
T61204500ML 500 mL
T612044L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. T61204100ML Supplier Sigma Aldrich Supplier No. T61204100ML
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as a reducing agent; can react with electrophiles to form phosphonates or phosphates; forms a stable complex with copper(I) iodide.

Specifications

CAS 867-13-0
Density 1.13 g/mL at 25°C
Boiling Point 142°C to 145°C (9 mmHg, lit.)
Molecular Formula C8H17O5P
Refractive Index n20/D 1.413
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
MDL Number MFCD00009177
Quantity 100 mL
Synonym P(EtO)3; P(OEt)3; P(EtO)3; P(OEt)3
Molecular Weight (g/mol) 224.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.