Learn More
MilliporeSigma™ Accessories for All-glass Filter Holder Kits, 90 mm
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | 90 mm all-glass filter holder |
| Material | Anodized Aluminum |
| Sterility | Non-sterile |
| Type | Spring Clamp, Aluminum |
| Diameter (Metric) Inside | 90 mm |