Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonitrilic chloride trimer, 98%

Catalog No. AC174280050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174280050 5 g Glass bottle
AC174280250 25 g Glass bottle
AC174281000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC174280050 Supplier Thermo Scientific Chemicals Supplier No. 174280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 940-71-6 | Cl6N3P3 | 347.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.65
MDL Number MFCD00006474
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Melting Point 111.0°C to 115.0°C
Quantity 5 g
Assay Percent Range 98%
Fieser 02,206; 04,386; 06,469
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether, benzene, toluene
Formula Weight 347.65
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 1.98
Density 1.9800g/mL
Chemical Name or Material Phosphonitrilic chloride trimer
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 213-376-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.