Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylitol, 99+%

Catalog No. AC225981000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225981000 100 g Plastic bottle
AC225980250 25 g Plastic bottle
AC225985000 500 g Plastic bottle
AC225980025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC225981000 Supplier Thermo Scientific Chemicals Supplier No. 225981000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 215.0°C to 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. Other polyols (GC)
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]3CH2OH
Quantity 100 g
Beilstein 01, 531
Merck Index 15, 10283
Solubility Information Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
Formula Weight 152.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Xylitol, 99+%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 201-788-0
RTECSNumber ZF0800000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.