Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Isoascorbic Acid, Sodium Salt 98%, Thermo Scientific™

Catalog No. AC275915000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC275915000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC275915000 Supplier Thermo Scientific Chemicals Supplier No. 275915000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-77-7
Molecular Formula C6H8NaO6
Molecular Weight (g/mol) 199.114
InChI Key WTTKTIBMFBIUSE-RKJRWTFHSA-N
Synonym sodium erythorbate
PubChem CID 66952495
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;sodium
SMILES C(C(C1C(=C(C(=O)O1)O)O)O)O.[Na]

Specifications

Melting Point 168°C to 172°C
Color White
pH 5.5 to 8 (5% aq. soln.)
Infrared Spectrum Authentic
Quantity 500 g
Merck Index 14, 5126
Specific Rotation +95.5° to +98° (25°C, 589nm) (c=10, water)
Formula Weight 198.11
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Isoascorbic acid, sodium salt, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.