Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 12, 66% Di-Lithium Tetraborate/34% Lithium Metaborate, Spectromelt™, MilliporeSigma™

Catalog No. M1118025000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1118025000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1118025000 Supplier MilliporeSigma Supplier No. 1118025000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12007-60-2 , 13453-69-5
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

CAS Max % 70.0
Physical Form Solid
Quantity 5 kg
Assay Percent Range 66% Di-Lithium Tetraborate/34% Lithium Metaborate
Odor Odorless
Packaging Plastic bottle
Chemical Name or Material A 12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.