CAS RN 12007-60-2
Lithium tetraborate, 98+%, for analysis
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Lithium tetraborate, 99.6% (metals basis)
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
di-Lithium tetraborate, MilliporeSigma™
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N Synonym: di-Lithium tetraborate IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Spectromelt A 10, MilliporeSigma™
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N Synonym: lithium tetraborate, di-lithium tetraborate IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Lithium tetraborate, 99% (metals basis)
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Lithium Tetraborate, Solstice
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Sigma Aldrich Lithium tetraborate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
A 12, 66% Di-Lithium Tetraborate/34% Lithium Metaborate, Spectromelt™, MilliporeSigma™
CAS: 12007-60-2 Molecular Formula: B4Li2O7 Molecular Weight (g/mol): 169.11 MDL Number: MFCD00011083 InChI Key: RDZCISUGRQUANA-UHFFFAOYSA-N IUPAC Name: dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2