Learn More
Eurofins Discoverx A-1210477, 5 mg. (92-1309)
Supplier:  Eurofins Discoverx 921309
Encompass_Preferred
A-1210477 is a small molecule MCL-1 inhibitor with sufficient potency to disrupt the tight-binding MCL-1-BIM complex in living cells, achieving the hallmarks of intrinsic apoptosis, and demonstrating clear-on-target cancer cell killing capability either as a single agent or in combination with ABT-263 (Navitoclax). Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.