Learn More
Selleck Chemical LLC A-485 2mg 1889279-16-6
Supplier: Selleck Chemical LLC S87402MG
A-485 is a potent, selective and drug-like p300/CBP catalytic inhibitor with an IC50 of 0.06 ?M for p300 HAT. It is selective over BET bromodomain proteins and >150 non-epigenetic targets. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.