Learn More
A-549 (human lung carcinoma) Nuclear extract lysate, Denatured; Abnova
Shop All Abnova Corporation ProductsDescription
Western Blotting
Specifications
Specifications
| Description | Nuclear extract cell lysate (denatured) |
| Lysis Buffer | Buffer A: 10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT. Buffer C: 20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. Buffer D : 20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. |
| Preparation Method | Nuclear extract was prepared by using a modified protocol of Dignam et al. Cells were Harvested and homogenized in Buffer A, and then centrifugated at 25,000 g for 20 minutes to remove cytoplasm and pellet the nuclei. The pellet was re-suspended in Buffer C, and then the suspensions were centrifuged to collect nuclear extract. The supernatant was dialyzed against Buffer D. The dialysate was then centrifuged, divided into aliquots, and stored at -80°C. The protein concentration was determined by the method of Bradford (Bio-Rad protein assay, microplate standard assay). The lysate was adjusted to 2.5 mg/ml, and then mixed with 5X Sample Buffer to become final 2 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 50 μg |
| Storage Buffer | In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). |
| Host Species | Human |
| Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| For Use With (Application) | Western Blot |
| Tissue | Lung |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.