Learn More
A-549 (human lung carcinoma) Whole cell lysate, Non-denatured, Abnova
Shop All Abnova Corporation ProductsDescription
- Tissue: Lung
- Host: Human
- Lysis Buffer: Modified RIPA Lysis Buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA Lysis Buffer
Western Blotting, Immunoprecipitation
Specifications
Specifications
| Description | Whole cell lysate (non-denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 200 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.