Learn More
Eurofins Discoverx A-61603 hydrobromide, 10 mg. (92-1230)
Supplier:  Eurofins Discoverx 921230
Encompass_Preferred
A 61603 is a synthetic small molecule and potent agonist of the G protein-coupled alpha 1A receptor (α1A), at least 35-fold selective vs α1B or α1D. More potent stimulant of phosphoinositide hydrolysis than norepinephrine, and inhibited by prazosin. Key role in modulation of the sympathetic nervous system, and increases spontaneous calcium transients more potently than phenylephrine. May be implicated in diseases such as hypertension. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9753
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.