Learn More
Eurofins Discoverx A-68930, 10 mg. (92-1043)
Supplier:  Eurofins Discoverx 921043
Encompass_Preferred
A68930 is a synthetic molecule developed by Abbott that is selective for D1-like dopamine receptor over D2-like (EC50=2.1 and 3910 nM, respectively). Produces antidepressant and anorectic effects in animals. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9588
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.