Learn More
AG Scientific Inc A 83-01, 1 MG
Supplier:  AG Scientific Inc A17641MG
Encompass_Preferred
A83-01 is a selective inhibitor of TGF-type I receptor ALK5 kinase, type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 (IC values are 12, 45 and 7.5 nM respectively). Blocks phosphorylation of Smad2 and inhibits TGF-induced epithelial-to-mesenchymal transition. Helps maintain homogeneity and long-term in vitro self-renewal of human induced pluripotent stem cells (iPSCs)Not for human therapeutic use or for medicinal purposes.CAS Number: 909910-43-6Molecular Weight: 421.52Chemical Formula: C25H19N5SSolubility: DMSO (50 mM)Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.