Learn More
BIOGEMS INTERNATIONAL INC A 83-01 25mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 909436025MG
Encompass_Preferred
A 83-01 is a potent activing receptor-like kinase inhibitor (ALK4 IC50 45nM, ALK5 IC50 12nM, ALK7 IC50 7.5nM). It is a more potent inhibitor of ALK5 than SB 431542 and is reported to inhibit Smad signaling and epithelial-to-mesenchymal transition by transforming growth factor-beta. However, A 83-01 had little or no effect on bone morphogenetic protein type I receptors, p38 mitogen-activated protein kinase, or extracellular regulated kinase. It is used to inhibit differentiation in iPSCs and maintain in vitro cellular self-renewal.
Catalog No. 50-153-0534
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.