Learn More
Apexbio Technology LLC A-966492, 5mg. Cas: 934162-61-5 MFCD: MFCD17215207. PARP-1/-2 inhibitor, highly potent
SDP
Supplier:  Apexbio Technology LLC A41605
Encompass_Preferred
A-966492 is an inhibitor of PARP-1 with Ki value of 1nM [1].PARP-1 belongs to the poly(ADP-ribose) polymerases (PARPs) family, it contributes to the resistance happened after cancer therapy. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4896
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.