Learn More
ATCC ATCC Quantitative Genomic RNA, Influenza A (H1N1) strain A/Baltimore/JH-22377/2022 (VR-3403D)
Supplier:  ATCC VR3403D
Encompass
Quantitative Genomic RNA from Influenza A virus (H1N1) strain A/Baltimore/JH-22377/2022 is a precisely quantified RNA standard intended for molecular assay development, verification, validation, and routine monitoring of diagnostic performance. The material is designed for use as a qPCR standard and other nucleic acid-based applications.
  • Quantified RNA standard suitable for generating qPCR standard curves.
  • Intended for assay development, verification, and validation.
  • Supports monitoring of day-to-day test variation and lot-to-lot performance.
  • Genome-sequenced strain for reliable molecular analyses.
  • Supplied frozen in a 100 μL aliquot; store at -70°C or colder.
Catalog No. NC3760734