Learn More
Cytiva (Formerly Pall Lab) A/D Glass Fiber Filter, 8 x 10 in. (25/pkg)
Supplier:  Cytiva 66227
Encompass_Preferred
Superior Grade Filters for a Variety of Biological and Environmental Methods-Type A/E meets the requirements for suspended solids testing, as described in Standard Methods for the Examination of Water and Wastewater, current edition.-Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions.-Extends filter life and reduces final filter changes with high capacity prefilters.-Eliminates sample contamination. Binder-free borosilicate glass fiber has no added extractables.-Filters a wide range of particulate loads and viscous solutions with a selection of filter thicknesses from which to choose.-Filters with binder offer excellent wet strength for easy handling and filter integrity. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.