Learn More
Medchemexpress LLC a-D-Glucosamine pent 10g | 7784-54-5 | 10 G

Supplier: Medchemexpress LLC HY11499810G
Alpha-D-glucosamine pentaacetate is the fully acetylated derivative of D-glucosamine used as an intermediate and reagent in glycobiology and carbohydrate chemistry. It is supplied as a solid for synthetic and analytical workflows, including derivatization and glycan synthesis.
- Fully acetylated amino sugar derivative for carbohydrate synthesis.
- High purity suitable for research (≥97.0%).
- Useful as a synthetic intermediate and for glycobiology studies.
- Solid form for convenient storage and handling.
- Molecular formula C16H23NO10; molecular weight 389.35 g·mol⁻¹; CAS 7784-54-5.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.