Learn More
ATCC Quantitative Genomic RNA, Influenza A (H3N2) strain A/Hong Kong/8/68 (100 μL, frozen)
Supplier: ATCC VR1679DQ
Quantitative genomic RNA isolated from Influenza A virus (H3N2) strain A/Hong Kong/8/68 provided as a frozen, quantitative preparation for assay development, verification, validation, and for generating qPCR standard curves to determine viral load.
- Prepared genomic RNA from Influenza A (H3N2), strain A/Hong Kong/8/68
- Quantitative format suitable for generating qPCR standard curves
- Packaged as a 100 μL frozen aliquot
- Store at -70°C or colder
- Intended for laboratory research use only