Learn More
ABclonal Technology A0052-500ul MGMT
SDP
Supplier:  ABclonal Technology A0052500UL
Encompass_Preferred
Alkylating agents are potent carcinogens that can result in cell death mutation and cancer The protein encoded by this gene is a DNA repair protein that is involved in cellular defense against mutagenesis and toxicity from alkylating agents The protein catalyzes transfer of methyl groups from O(6)-alkylguanine and other methylated moieties of the DNA to its own molecule which repairs the toxic lesions Methylation of the genes promoter has been associated with several cancer types including colorectal cancer lung cancer lymphoma and glioblastoma
Catalog No. 50-001-36582
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.