Learn More
TARGETMOL CHEMICALS INC A09-003 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T794041MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. A09-003 is a novel cell cycle protein-dependent kinase-9 CDK-9 inhibitor.A09-003 inhibits the proliferation of a variety of leukemia cell lines and inhibits the increase of leukemia sequence-1 protein in myeloid cells.A09-003 also induces apoptosis, decreases RNA polymerase II activity, and decreases Mcl-1 expression. Purity 99.84%
Catalog No. 50-287-6780
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.