Learn More
ABclonal Technology A10277-1000ul KCNMB2
Supplier: ABclonal Technology A102771000UL
MaxiK channels are large conductance voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability MaxiK channels can be formed by 2 subunits the pore-forming alpha subunit and the modulatory beta subunit The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents Alternative splicing results in multiple transcript variants of this gene Additional variants are discussed in the literature but their full length nature has not been described
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.