Learn More
ATCC A2058; Melanoma; Human (Homo sapiens)
Supplier:  ATCC CRL3601
Encompass
A2058 is a cell exhibiting epithelial morphology isolated from the skin of a White, 43-year-old male patient with melanoma. This product is an ATCC manufactured and accessioned progeny of CRL-11147.
Catalog No. NC2470686
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.