Learn More
ABclonal Technology A18199-1000ul K33-linkage Poly
SDP
Supplier:  ABclonal Technology A181991000UL
Encompass_Preferred
This gene encodes ubiquitin one of the most conserved proteins known Ubiquitin has a major role in targeting cellular proteins for degradation by the 26S proteosome It is also involved in the maintenance of chromatin structure the regulation of gene expression and the stress response Ubiquitin is synthesized as a precursor protein consisting of either polyubiquitin chains or a single ubiquitin moiety fused to an unrelated protein This gene consists of three direct repeats of the ubiquitin coding sequence with no spacer sequence Consequently the protein is expressed as a polyubiquitin precursor with a final amino acid after the last repeat An aberrant form of this protein has been detected in patients with Alzheimers disease and Down syndrome Pseudogenes of this gene are located on chromosomes 1 2 13 and 17 Alternative splicing results in multiple transcript variants
Catalog No. 50-001-10737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.