Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGuard™ A21A5 Series Powder-Free Vinyl Stretch 4.3 mil Exam Gloves, Cream

Catalog No. 17004383
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-383 Small
17-004-384 Medium
17-004-385 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-004-383 Supplier VGuard™ Supplier No. A21A51
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cream A21 Vinyl Stretch Exam Gloves resist abrasions, cuts and punctures while offering a comfortable fit and feel.

  • Vinyl
  • Powder Free
  • Non-Sterile
  • Ambidextrous
  • Exam Grade
  • Color: Cream
  • Texture: Smooth
  • Palm Thickness: 4.3 mil
  • Fingertip Thickness: 4.3 mil
  • Length: Standard (23 cm)
  • Cuff: Beaded
  • FDA Compliant.
  • 100 Gloves/Box

Specifications

Chlorinated Yes
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Cream
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 4.3 mil
Latex Free Yes
Material Vinyl
Palm Thickness (English) 4.3 mil
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (English) 4.3 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model A21A51
Series Vanguard Safety
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.