Learn More
ABclonal Technology A1631-100ul FSP1/S100A4
SDP
Supplier:  ABclonal Technology A1631100UL
Encompass_Preferred
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation S100 genes include at least 13 members which are located as a cluster on chromosome 1q21 This protein may function in motility invasion and tubulin polymerization Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis Multiple alternatively spliced variants encoding the same protein have been identified
Catalog No. 50-001-10019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.