Learn More
Description
- Allows for maximum productivity and safety for you and your employees
- The material is strong and abrasion resistant, and filters out many water-based liquids and dry particles
- Includes seamless front (for primary exposure area), a 1 inch zipper flap, hood, elastic wrists and new skid-resistant boots that provide up to 68% improvement in traction on wet and dry surfaces when compared to our former boot material
- Protect against hazardous dry particles, aerosols and non-hazardous light liquid splashes, and they're perfect for environments such as pharmaceutical research, aerospace, maintenance and general manufacturing
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Color | White |
| Disposable | Single-use |
| Cuff Style | Elastic |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Coating | Anti-static |
| Back Style | Elastic |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.