Learn More
Ansell KleenGuard™ A30 Coveralls, Attached Hood and Boots
Catalog No. 19005103 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-103 8X-Large
19-005-094 Small
19-005-095 Medium
19-005-096 Large
19-005-097 X-Large
19-005-098 2X-Large
19-005-099 3X-Large
19-005-100 4X-Large
19-005-101 5X-Large
19-005-102 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-005-103 missing translation for 'mfr' Ansell missing translation for 'supplierNo' 48970
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Coveralls feature breathable fabric and Reflex design, which offers excellent fit and freedom of movement.
  • Allows for maximum productivity and safety for you and your employees
  • The material is strong and abrasion resistant, and filters out many water-based liquids and dry particles
  • Includes seamless front (for primary exposure area), a 1 inch zipper flap, hood, elastic wrists and new skid-resistant boots that provide up to 68% improvement in traction on wet and dry surfaces when compared to our former boot material
  • Protect against hazardous dry particles, aerosols and non-hazardous light liquid splashes, and they're perfect for environments such as pharmaceutical research, aerospace, maintenance and general manufacturing
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 8X-Large
Gender Unisex
Color White
Disposable Single-use
Cuff Style Elastic
Closure Type Front Zipper
Sterility Non-sterile
Coating Anti-static
Back Style Elastic
Certifications/Compliance NFPA 99 Compliant