Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGuard™ A31A1 Series Powder-Free Latex 5.5 mil Exam Gloves, Cream

Catalog No. 17004405
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-405 Small
17-004-406 Medium
17-004-408 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-004-405 Supplier VGuard™ Supplier No. A31A11
Add to Cart
Add to Cart
This item is not returnable. View return policy

A31 Cream Latex Exam Gloves provide all purpose protection for medical use. Latex material fits around hand for superior comfort and dexterity.

  • Latex
  • Powder-free
  • Non-Sterile
  • Ambidextrous
  • Exam Grade
  • Color: Cream
  • Texture: Finger and Palm
  • Palm Thickness: 4.5 mil
  • Fingertip Thickness: 5.5 mil
  • Length: Standard (23 cm)
  • Cuff: Beaded
  • 100 Gloves/Box

Specifications

Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Cream
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 5.5 mil
Latex Free No
Material Latex
Palm Thickness (English) 4.5 mil
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (English) 5.5 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model A31A11
Series Vanguard Safety
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.