Learn More
Biolegend Alexa Fluor 488 anti-human CD45
Supplier:  Biolegend 304019
Encompass_Preferred
Alexa Fluor 488 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-166-060