Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A4GNT Monoclonal Antibody (OTI1F7)
GREENER_CHOICE

Catalog No. PIMA526434
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA526434 100 μL
1 options

Catalog No. PIMA526434

Supplier: Invitrogen™ MA526434

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A4GNT Monoclonal Antibody for Western Blot

This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A4GNT
Applications Western Blot
Classification Monoclonal
Clone OTI1F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene A4GNT
Gene Accession No. Q14BT6, Q9UNA3
Gene Alias A4GNT; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; AV080780; EGK_12072; Gm798; GnTalpha4; MGC149493
Gene Symbols A4GNT
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 121-340 of A4GNT produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333424, 485683, 51146, 685758
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.