Learn More
Biolegend Alexa Fluor 647 anti-ERK1/2 Phospho (Thr202/Tyr204)
Supplier:  Biolegend 675503
Encompass_Preferred
Alexa Fluor 647 anti-ERK1/2 Phospho (Thr202/Tyr204) [4B11B69]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-7343
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.