Learn More
Biolegend Alexa Fluor 647 anti-rat CD45
Supplier:  Biolegend 202211
Encompass_Preferred
Alexa Fluor 647 anti-rat CD45 [OX-1]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 25 ug
Catalog No. 50-165-369
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.