Learn More
ABclonal Technology A9151-1000ul 14-3-3 alpha/beta
SDP
Supplier:  ABclonal Technology A91511000UL
Encompass_Preferred
This gene encodes a protein belonging to the 14-3-3 family of proteins members of which mediate signal transduction by binding to phosphoserine-containing proteins This highly conserved protein family is found in both plants and mammals The encoded protein has been shown to interact with RAF1 and CDC25 phosphatases suggesting that it may play a role in linking mitogenic signaling and the cell cycle machinery Two transcript variants which encode the same protein have been identified for this gene
Catalog No. 50-001-36797
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.