Learn More
ABclonal Technology A9738-1000ul Collagen VI/COL6A
SDP
Supplier:  ABclonal Technology A97381000UL
Encompass_Preferred
The collagens are a superfamily of proteins that play a role in maintaining the integrity of various tissues Collagens are extracellular matrix proteins and have a triple-helical domain as their common structural element Collagen VI is a major structural component of microfibrils The basic structural unit of collagen VI is a heterotrimer of the alpha1(VI) alpha2(VI) and alpha3(VI) chains The alpha2(VI) and alpha3(VI) chains are encoded by the COL6A2 and COL6A3 genes respectively The protein encoded by this gene is the alpha 1 subunit of type VI collagen (alpha1(VI) chain) Mutations in the genes that code for the collagen VI subunits result in the autosomal dominant disorder Bethlem myopathy
Catalog No. 50-001-36058
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.