Learn More
Medchemexpress LLC HY-124293 5mg Medchemexpress, AA147 CAS:393121-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1242935MG
Encompass_Preferred
Medchemexpress, HY-124293 5mg AA147 CAS:393121-74-9 AA147, a small molecule endoplasmic reticulum (ER) proteostasis regulator, selectively activates ATF6 arm of the unfolded protein response (UPR) extracted from patent WO2017117430A1, compound 147*. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9987
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.